Honeywell PBI Aluminized LTO Proximity Pants
For applications where sustained operations in high heat are anticipated
Manufacturer: Total Fire Group SAEGSA00478 3XL
 View more versions of this product
Catalog No. 19-165-763
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Size 3X-Large
    Material Aluminized PBI Silver; Two-layer Spunlace/meta-aramid Thermal Liner; CrossTech™/Nomex™ Moisture Barrier
    Color Silver
  • Description & Specifications
Related Products
Product Content Feedback