Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell PBI Bronze Aramid Fiber LTO Fire Protective Garment

Lightweight, one-piece FPG ensemble with snap-in removable liner

$728.16 - $1322.19

Specifications

Color Bronze
View More Specs

Products 4
Catalog Number Mfr. No. Material Size Length (Metric) Style Price Quantity & Availability  
Catalog Number Mfr. No. Material Size Length (Metric) Style Price Quantity & Availability  
19-821-048
View Documents
Ergodyne
17866
Bronze PBI/Dupont™ Kevlar™ aramid fiber Outer; Nomex Thermal Liner Large 89 cm Regular inseam
Case of 144 for $1,322.19
Add to cart
 
19-821-042
View Documents
Ergodyne
17855
Bronze PBI/Dupont™ Kevlar™ aramid fiber Outer; Nomex Thermal Liner Small 83.8 cm Regular inseam
Case of 144 for $728.16
Add to cart
 
19-821-045
View Documents
Ergodyne
17863
Bronze PBI/Dupont™ Kevlar™ aramid fiber Outer; Nomex Thermal Liner Medium 87.6 cm Regular inseam
Case of 144 for $1,322.19
Add to cart
 
19-821-051
View Documents
Ergodyne
17874
Bronze PBI/Dupont™ Kevlar™ aramid fiber Outer; Nomex Thermal Liner X-Large 90.1 cm Regular inseam
Case of 144 for $1,322.19
Add to cart
 
Description

Description

  • Outer shell of bronze 7.5 oz. Pbi™/Kevlar™ aramid fibers ripstop fabric with enhanced water repellency
  • Thermal liner of iced blue 7.5 oz. Nomex™ Semi-slick: 3.5 oz. filament/spun Nomex facecloth, and 4.0 oz. Nomex/Kevlar™ aramid fibers batt
  • Moisture barrier of 5 oz. Crosstech™ with Nomex facecloth: 1.7 oz. Crosstech selectively permeable film laminated to 3.3 oz. Nomex pajama check woven substrate
  • Raglan sleeves for unrestricted movement, plus front zipper running from neck to crotch for quick donning/doffing
  • Stand-up collar with Velcro™ closure
  • Front zipper with storm flap and Velcro closure
  • Elasticized waist for comfortable fit; articulating rapid-rescue strap
  • Double-stitched, lime Scotchlite™ 2 in.-wide reflective trim at chest, hip, wrist, and ankle

  • Reinforced elbows
  • Elasticized cuff with Velcro closure and over-the-thumb Kevlar™ aramid fibers wristlets
  • Two double-stitched 8 x 7.5 x 2.5 in. bellows pockets with reinforced bottoms and 5 in. flap
  • Double-stitched cushioned knees with 11 x 9 in. gray Arashield™
  • Double-stitched pant cuffs with gray Arashield
  • Zippered boot access panel on each leg

Order Info

Available through Fisher Safety's GSA Advantage!™ Schedule 84.

Certifications

Approved and certified for use onboard U.S. Navy ships and submarines per NAVSEA 05P14.

Compliance

Meets NFPA 1971, 2007 Edition.

Warranty and Services

Lifetime guarantee against defects in materials or workmanship

Specifications

Specifications

Bronze
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit