Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Perfect-Coat Cut-Resistant Gloves

Catalog Number p-4414164
Click to view available options
Size:
Small
Medium
Large
X-Large
Hem Color:
Blue
Red
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Coating Material Natural Rubber
Color Yellow
Cut Resistance Rating ANSI A4
Material Aramid Fiber, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Protects against cuts and abrasions from glass, metals and other sharp objects
Glove Cuff Color Red
Glove Cuff Style Knit
Hem Color Red
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit