Learn More
Honeywell Perfect-Coat™ Rubber Coated Gloves
Ergonomic design conforms snugly to the natural shape of the hand, reducing hand fatigue
Manufacturer:  Honeywell 300PCS
 View more versions of this product
Catalog No. 18-999-4410
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Specifications Description & Specifications
Specifications
Palm, finger
Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
Knit
Rubber, natural
Small
Red
Level 1
Rough
construction, recycling, lumber and mill work, Glass handling, Metal stamping, automotive, landscaping
Cotton, Polyester
Blue,Gray