Honeywell Perfect-Coat™ Rubber Coated Gloves

Ergonomic design conforms snugly to the natural shape of the hand, reducing hand fatigue

Manufacturer: Honeywell 300PCM

 View more versions of this product

Catalog No. 18-999-4410A

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


    Specifications

    Cleaning Procedure Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
    Exterior Finish Rough
    Glove Cuff Style Knit
    Glove Cuff Color Blue
    For Use With (Application) construction, recycling, lumber and mill work, Glass handling, Metal stamping, automotive, landscaping
    Material Natural Rubber
    Liner Material Cotton, Polyester
    Size Medium
    Coated Area Palm, finger
    Color Blue, gray
    Cut Resistance Rating Level 1

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit