Learn More
Honeywell Perfect-Coat™ Rubber Coated Thermal Gloves
Heavyweight brushed thermal cotton/poly liner adds warmth and comfort
Manufacturer:  Honeywell 451PCXL
 View more versions of this product
Catalog No. 19-014-388D
  Check Availability
Add to cart
Alert:
Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Specifications Description & Specifications
Specifications
Palm, finger
Rough
construction, recycling, lumber and mill work, Glass handling, Metal stamping, automotive, landscaping
Cotton, Polyester
Gray,Light Gray
Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
Knit
Rubber, natural
X-Large
White