Honeywell Perfect Fit™ Cut-Resistant Gloves

Dyneema fiber gloves provide a high level of cut protection in a lightweight glove

Manufacturer: Honeywell PF13XL

 View more versions of this product

Catalog No. 19-049-895E

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Cleaning Procedure Withstands laundry room Caustics up to pH 15 and Water temperatures up to 180°C, they may be repeatedly Bleached or disinfected without harm and will not discolor
    Hand Ambidextrous
    Hem Color Green
    For Use With (Application) food processing, food packaging, meat and poultry industries, restaurants, commercial fishing and canning
    Material HPPE
    Size X-Large
    Style 13-Cut
    Weight Light weight
    Color White with green cuff
    Cut Resistance Rating Level 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit