Learn More
Honeywell™ Perfect Fit™ Cut-Resistant Gloves
Dyneema fiber gloves provide a high level of cut protection in a lightweight glove
Manufacturer:  Honeywell™ HPF7M/USE19816646
 View more versions of this product
Catalog No. 19-049-893C
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Lightweight, low-linting and does not easily absord liquids; these gloves are excellent for food processing applications
  • Color-coded PowerCuff™ helps to eliminate cuff “blow-out” from repeated laundering
  • Can easily be sorted by size
  • Sold individually
  • Complies with USDA and FDA regulations, 21 CFR, for use in food processing
Specifications
Specifications
Safety
Withstands commercial laundry-room detergents (up to pH 15) and cool water temperatures up to 49°C (120°F). Lower heat settings
Blue
HPPE
7-Cut
Yellow
ASTM
Level 5
Ambidextrous
food processing, food packaging, meat and poultry industries, restaurants, commercial fishing and canning
Medium
Heavy weight
Yellow