Learn More
Honeywell™ Perfect Fit™ Cut-Resistant Gloves
Dyneema fiber gloves provide a high level of cut protection in a lightweight glove
Manufacturer:  Honeywell™ HPF7L
 View more versions of this product
Catalog No. 19-049-893D
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Lightweight, low-linting and does not easily absord liquids; these gloves are excellent for food processing applications
  • Color-coded PowerCuff™ helps to eliminate cuff “blow-out” from repeated laundering
  • Can easily be sorted by size
  • Sold individually
  • Complies with USDA and FDA regulations, 21 CFR, for use in food processing
Specifications
Specifications
Safety
Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
Yellow
HPPE
7-Cut
Blue
ASTM
Level 5
Ambidextrous
food processing, food packaging, meat and poultry industries, restaurants, commercial fishing and canning
Large
Heavy weight
Blue