Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Pure Fit PF570 HPPE Cut-Resistant Gloves, Nitrile Coated

Catalog Number 191305800
Click to view available options
Size:
Small
Medium
Large
Glove Cuff Color:
Blue
Red
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Nitrile coated ANSI Cut Level 4 protection gloves offer dexterity and tactile sensitivity for work requiring precision handling. Seamless knit construction eliminates uncomfortable seams that can rub or chafe.

  • White high performance polyethylene (HPPE) and stainless steel blended shell
  • Lycra™ plating provides comfortable, snug fit
  • Gray nitrile palm and fingertip coating provides extra durability and secure grip
  • 13-gauge, lightweight construction
TRUSTED_SUSTAINABILITY
ANSI Abrasion Rating ANSI 4
Coating Material Nitrile
Color Light Gray, Gray
Cut Resistance Rating ANSI A5
Material HPPE, Stainless Steel Fiber
Product Type Cut-resistant Gloves
Size Medium
EN388 Standard EN 388
For Use With (Application) Precision handling
Glove Cuff Color Blue
Hem Color Blue
Model PF570-M
Series Pure Fit PF570
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit