Honeywell PowerCoat™ Disposable Tri-Polymer Gloves

Manufactured for comfort and safety

Manufacturer: Honeywell PSDTRIPS

 View more versions of this product

Catalog No. 19-130-1847A

  Check Availability
Add to Cart

Alert

Product contains latex which may cause a reaction in some individuals.


    Specifications

    Exterior Finish Rough
    Glove Cuff Style Rolled
    For Use With (Application) food processing, chemical, biohazards, mail sorting and medical
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Nitrile
    Powdered No
    Size Small
    Color Purple

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit