Learn More
Honeywell™ PowerCoat™ Double Dipped PVC Gloves
Super flexible PVC coating resists caustics, bases, alcohols and many acids
Manufacturer:  Honeywell™ 620L
 View more versions of this product
Catalog No. 19-049-925B
Add to cart
Description
Description
  • Superior grip and dexterity, flexibility reduces hand fatigue and promotes increased productivity
  • Supple as natural latex, but has the chemical resistance of PVC
  • Ideal alternative for workers with latex allergies
  • Excellent for multiple applications including glass handling, injection molding, plastic processing and more
Order Info
Available in packs of 12 pairs.
Specifications
Specifications
Safety
Pinked
PVC
620
12 in.
Rough
Cotton
Large
Orange
30.5 cm