Honeywell PowerCoat Flock-Lined Nitrile Gloves
Provide excellent chemical resistance, flexibility and dexterity
Manufacturer: Honeywell 772-M
 View more versions of this product
Catalog No. 19-130-1806B
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications ANSI/ISEA 105-2005, CE, EN 388, EN 374-3-2003, ASTM
    Glove Cuff Style Elastic
    For Use With (Application) Oily greasy applications, agricultural spraying, municipal Water treatment, animal husbandry, protection from fuels, sewer and tank cleaning, refineries, chemical splash protection, painting
    Size Medium
    Length (English) 26 in.
    Length (Metric) 66cm
    Lining Cotton, Polyester
    Material Nitrile
    Coating Material Rough
    Color Yellow
  • Description & Specifications
Related Products
Product Content Feedback