Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ PowerCoat™ Nitrile Gloves

Catalog Number p-3752812
Click to view available options
Size:
Small
This item is not returnable. View return policy
This item is not returnable. View return policy

PowerCoat™ 12 in. disposable nitrile gloves offer chemical and liquid protection in a variety of industrial environments.

  • Disposable nitrile
  • Latex-free
  • 12 inch length
  • Rolled cuff
  • 8 mil
  • Ideal for agriculture, chemical and oil/gas industries
TRUSTED_SUSTAINABILITY
Certifications/Compliance ANSI
Chemotherapeutic Rated No
Applications Industrial
Color Blue
Disposable Single-use
Exterior Finish No Texture
Laboratory/Cleanroom Use No
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 8 mil
Thickness (Metric) 0.20 mm
Model PSD-NI8-S
Series PowerCoat™
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit