Honeywell Premium Natural Rubber Latex Gloves

Manufacturer: Honeywell PNLB1815/O/11

 View more versions of this product

Catalog No. 19-150-1077

  Check Availability
Add to Cart

    Specifications

    Certifications 21 CFR, ANSI, ISEA 105, ASTM D 3389-05, F1790-97, USDA, FDA
    Exterior Finish Embossed, Diamond
    Glove Cuff Style Beaded
    For Use With (Application) automotive assembly, chemical handling and processing, food processing and preparation, janitorial
    Length (Metric) 38.1cm
    Length (English) 15 in.
    Material Natural Rubber
    Powdered No
    Size 11
    Thickness 18mil
    Color Orange, yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit