Learn More
Honeywell™ PRO 3009 Leather Boots
CrossTech™ fabric bootie system laminated to quilted, fungus-resistant Cambrelle™ and Kevlar™ felt linings
Manufacturer:  Honeywell™ BT30091051
 View more versions of this product
Catalog No. 19-814-359
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Corrosion-resistant steel ladder shank and puncture-resistant bottom plate
  • Double-stitched, reinforced aggressive lug soles
  • 14 in. high black, pull-on bunker style boot with side and back stays made from top-grade Military AB water-resistant leather
  • Oblique safety-toe cap
  • Slip-resistant tread design
  • Full wraparound Goodyear™ Welt
†“D” width. Additional widths available through your Fisher Scientific™ Customer Service Representative.
Compliance
Meet NFPA 1971, 2007 Edition requirements.
Specifications
Specifications
14 in.
Leather, CrossTech
10.5C