Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ PRO 3009 Leather Boots, Wide Width
Engineered for comfort, safety and control
Supplier: Honeywell™ BT30091003
Description
- 14 in. pull-on structural boot
- Goodyear welt construction
- Vibram™ lug soles
- Premium full-grain military AB leather
- Full Crosstech™ moisture barrier bootie with 7.5 oz. Kevlar™ felt thermal barrier and Cambrelle™ liner
- Silicone impregnated leather toe cap
- Oblique steel toe, steel bottom plate and shank
- Thermoplastic heel counter
- APT barcoded
- Widths: E, EE, EEE
- Half sizes: 5.5 to 12.5
- Full sizes: 5 to16
Certifications
NFPA 1971
Specifications
14 in. | |
Leather | |
10E |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction