Honeywell PRO 5006 Structural Firefighting Boots, D Width Available on GSA/VA Contract for Federal Government customers only.
Superior combination of comfort and absorbency
Manufacturer: Total Fire Group BT5006B0752
 View all options for this product
Catalog No. 19-167-401
  Check Availability
Add to Cart
    Height (English) 14 in.
    Material Pro-V Liner
    Size 7.5D
    Quilted to 7.5 oz Kevlar™ felt thermal barrier and laminated to fully encapsulating Crosstech™ Footwear Fabric bootie. Crosstech™ moisture barrier provides superior liquid penetration resistance to blood, body fluids and NFPA tested common chemicals.
    • Military AB leather construction insulates and adds abrasion-, puncture-, water- and chemical-resistance
    • With heavy duty 0.75 in. reinforced straps mounted to the exterior of boot and anchored with a 2 x 1.5 in. shield-shaped base
    • External texon board shinguard protects and guards sensitive shin area of legs
    • Patented PowerToe™ cover resists toe abrasion and provides unparalled wear over the lifetime of the boots
    • Oblique toe safety cap provides additional room in height and width of boot
    • Fire/Ice PowerSole™ angled lugs provide substantial edging and gripping capabilities
    • Molded thermoplastic heel counter locks foot into the boot and anchors it in place
    • Patented PowerHeel™ cover protects heel area from abusive doffing of boots
    • Corrison-resistant steel ladder shank and stainless steel, full length, toe-to-heel puncture resistant bottom plate
  • Description & Specifications
Related Products
Product Content Feedback