Honeywell PRO 5050 Struximity Leather Boots
Lightweight, comfortable, high-speed leather pull-on bunker boots dual certified for both Structural and Proximity (ARFF) firefighting
Manufacturer: Total Fire Group BT5050115D
 View more versions of this product
Catalog No. 19-814-823
  Check Availability
Add to Cart
    Specifications
    Size 11.5D
    Material Leather; CrossTech Fabric; Kevlar Felt Lining; Cambrelle Liner
    Weight (English) 6.6 lb.
    Certifications/Compliance Certified to NFPA 1971, current edition for Proximity and Structural Fire Fighting
  • Description & Specifications
Related Products
Product Content Feedback