Learn More
Honeywell™ PRO 6006 Technical Rescue Boots, EE Width
Provide light and flexible protection
Manufacturer:  Honeywell™ BT60061154
 View more versions of this product
Catalog No. 19-167-371
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This 100% waterproof, lace-to-toe boot features a reinforced stitched down sole with Fire/Ice compound. Vibram™ Fire/Ice sole improves diagonal and lateral movement as well as controlled acceleration in icy conditions.
  • Constructed of premium full grain military AB leather
  • Patented rubber metatarsal guard protects the five long bones in the forefoot
  • Composite toe cap eliminates hot/cold conductivity while protecting toes from impact injuries
  • Patented PowerToe™ improves the wearlife of boots
  • Wrap-around goodyear welt supports and reduces fatigue and ankle sprains and strains
  • Steel ladder shank and stainless steel bottom plate protects and supports the arch area of foot with heel to toe full coverage
  • Removable polyurethane insert dissipates fatigue for total comfort control
  • Barcoded for ease of recordkeeping responsibilities
  • Boots also include Fire Resistant Advance™ rip stop side panels, thermally bonded non-woven PRO-V liner, Crosstech™ moisture barrier and 1 in. cushion collar
Order Info
D and EEE widths also available.
Compliance
NFPA 1951, 2007 Edition Technical Rescue Recovery/Utility. NFPA 1999, 2003 Edition Emergency Medical Operations. NFPA 1992, 2005 Edition Liquid Splash. NFPA 1977, 2005 Edition Wildland (Optional).
Specifications
Specifications
8 in.
Leather, Pro-V Liner
11.5EE