Learn More
Honeywell™ Proximity 3-D Gloves
3-D pattern combines elk palm with aluminized back and sides of fingers
Manufacturer:  Honeywell™ GLBPRRWAXXXL
 View more versions of this product
Catalog No. 19-171-189H
Add to cart
Description
Description
  • 3-D pattern on elk leather palm with an aluminized Pbi™/Kevlar™ back and sides of fingers
  • Flex Bubbles at finger joints and Flex-tuck across carpal area for added comfort
  • CrossTech™ moisture barrier for higher breathability and heat resistance
  • Self-extinguishing fleece liner
  • Wing thumb
Certifications
USAF J-Fire tested and approved.
Compliance
Meet NFPA 1971, 2007 Edition requirements for proximity/ARFF operations. Meet bloodborne pathogen resistant (BPR) interface capabilities when worn with other BPR elements.
Specifications
Specifications
Wristlet
Self extinguishing Fleece
USAF J-fire tested
Dupont™ Kevlar™ aramid fibers, PBI™, aluminized
3X-Large