Learn More
Honeywell™ Proximity 3-D Gloves Available on GSA/VA Contract for Federal Government customers only.
3-D pattern combines elk palm with aluminized back and sides of fingers
Manufacturer:  Honeywell™ GLBPRRGA
 View more versions of this product
Catalog No. 19-823-791
Add to cart
Description
Description
  • 3-D pattern on elk leather palm with an aluminized Pbi™/Kevlar™ back and sides of fingers
  • Flex Bubbles at finger joints and Flex-tuck across carpal area for added comfort
  • CrossTech™ moisture barrier for higher breathability and heat resistance
  • Self-extinguishing fleece liner
  • Wing thumb
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Certifications
USAF J-Fire tested and approved.
Compliance
Meet NFPA 1971, 2007 Edition requirements for proximity/ARFF operations. Meet bloodborne pathogen resistant (BPR) interface capabilities when worn with other BPR elements.
Specifications
Specifications
Gauntlet
Self extinguishing Fleece
USAF J-fire tested
PBI™, aluminized, Dupont™ Kevlar™ aramid fibers
Large