Honeywell Proximity 3-D Gloves Available on GSA/VA Contract for Federal Government customers only.
3-D pattern combines elk palm with aluminized back and sides of fingers
Manufacturer: Total Fire Group GLBPRRGAXXXL
 View more versions of this product
Catalog No. 19-171-190H
  Check Availability
Add to Cart
    Specifications
    Glove Cuff Style Gauntlet
    Material Aluminized PBI™, Kevlar™
    Liner Material Self extinguishing Fleece
    Size 3X-Large
    Certifications USAF J-fire tested
    • 3-D pattern on elk leather palm with an aluminized Pbi™/Kevlar™ back and sides of fingers
    • Flex Bubbles at finger joints and Flex-tuck across carpal area for added comfort
    • CrossTech™ moisture barrier for higher breathability and heat resistance
    • Self-extinguishing fleece liner
    • Wing thumb
  • Description & Specifications
Related Products
Product Content Feedback