Learn More
Honeywell™ Proximity Gloves
Exceptionally comfortable firefighting glove
Manufacturer:  Honeywell™ GLBPRRWAMLS
 View more versions of this product
Catalog No. 19-171-191C
Add to cart
Description
Description
  • 3-D hand-shaped styling with staggered layer seaming
  • Self-shaped hand opening into fingers and glove body ease donning/doffing
  • Elk leather palm with Pbi™/Kevlar™ back and sides of fingers and wing thumb
  • Self-extinguishing fleece liner and Crosstech™ moisture barrier
  • Wristlet cuff with Wick-Guard stops water and fluid from reaching glove interior
  • Flex Bubbles at finger joints and Flex-Tuck across carpal area allow unrestricted movement
  • Regular cut with 4 in. military length knit wrist
Specifications
Specifications
Wristlet
Self extinguishing Fleece
USAF J-fire tested
Dupont™ Kevlar™ aramid fibers, PBI™, aluminized
Small