Honeywell Proximity Gloves Available on GSA/VA Contract for Federal Government customers only.
Exceptionally comfortable firefighting glove
Manufacturer: Total Fire Group GLBPRRWAMLXXXL
 View more versions of this product
Catalog No. 19-171-191H
  Check Availability
Add to Cart
    Specifications
    Glove Cuff Style Wristlet
    Material Aluminized PBI™, Kevlar™
    Liner Material Self extinguishing Fleece
    Size 3X-Large
    Certifications USAF J-fire tested
    • 3-D hand-shaped styling with staggered layer seaming
    • Self-shaped hand opening into fingers and glove body ease donning/doffing
    • Elk leather palm with Pbi™/Kevlar™ back and sides of fingers and wing thumb
    • Self-extinguishing fleece liner and Crosstech™ moisture barrier
    • Wristlet cuff with Wick-Guard stops water and fluid from reaching glove interior
    • Flex Bubbles at finger joints and Flex-Tuck across carpal area allow unrestricted movement
    • Regular cut with 4 in. military length knit wrist
  • Description & Specifications
Related Products
Product Content Feedback