Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Pure Fit™ Cut-Resistant Gloves

Ideal for cut and puncture protection without sacrificing dexterity

$26.62

Specifications

Coating Material Smooth
Color Black, Gray
Cut Resistance Rating Level 3
Material HPPE
For Use With (Application) Provides added grip, abrasion resistance and tactile sensitivity
View More Specs

Products 4
Catalog Number Mfr. No. Size Hem Color Price Quantity & Availability  
Catalog Number Mfr. No. Size Hem Color Price Quantity & Availability  
19-148-401
View Documents
Honeywell™
PF541M
Medium Blue
Pair for $26.62
Case of 144 PR for $2,840.60
Add to cart
 
19-148-400
View Documents
Honeywell™
PF541S
Small Red
Pair for $26.62
Case of 144 PR for $2,840.60
Add to cart
 
19-148-403
View Documents
Honeywell™
PF541XL
X-Large White
Pair for $26.62
Case of 144 PR for $2,840.60
Add to cart
 
19-148-402
View Documents
Honeywell™
PF541L
Large Yellow
Pair for $26.62
Case of 144 PR for $2,840.60
Add to cart
 
Description

Description

  • Shell made of low-linting, black and white speckled, Dyneema
  • Gray polyurethane smooth coating of palm and fingertip provides added grip, abrasion resistance and tactile sensitivity
  • Gray color of gloves hides dirt and grime
  • Available in different sizes

Specifications

Specifications

Smooth
Level 3
Provides added grip, abrasion resistance and tactile sensitivity
Light weight
Black, Gray
HPPE
Knit
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit