Learn More
Honeywell™ Pure Fit Gloves
Palm-dipped design reduces hand fatigue and offers extreme dexterity
Manufacturer:  Honeywell™ PF560M
 View more versions of this product
Catalog No. 19-825-204
Add to cart
Description
Description
  • Low-linting, lightweight nylon, seamless knit liners
  • Black nylon liner with black polyurethane coating (3/4 dipped to extend over knuckles)
  • Extra wear area provides longer wear and more comfort than cut and sewn gloves
  • Gauge: 13 cut
    • Certifications
      ISO 9001/2000
Specifications
Specifications
Full palm, fingertip
Smooth
small parts assembly, parts handling, auto repair, painting, hand and power tool operation
Nylon
Black
Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
Knit
Polyurethane
Medium
Blue