Honeywell Pure Fit Gloves

Palm-dipped design reduces hand fatigue and offers extreme dexterity

Manufacturer: Honeywell PF560S

 View more versions of this product

Catalog No. 19-825-203

  Check Availability
Add to Cart

    Specifications

    Cleaning Procedure Withstands commercial laundry room detergents (up to pH 14) and warm water temperatures up to 120°F. Lower heat settings up to 180°F are suggested for drying as extreme heat may cause shrinking and degradation.
    Exterior Finish Smooth
    Glove Cuff Style Knit
    Glove Cuff Color Red
    For Use With (Application) small parts assembly, parts handling, auto repair, painting, hand and power tool operation
    Material Polyurethane
    Liner Material Nylon
    Size Small
    Coated Area Full palm, fingertip
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit