Learn More
Honeywell™ Pure Fit™ Nitrile-Coated Dyneema Gloves
Lightweight, breathable fibers provide superior tactile sensitivity for precision handling and delicate operations
Manufacturer:  Honeywell™ PF545M
 View more versions of this product
Catalog No. 19-816-644
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Dyneema fiber provides cut and abrasion resistance in light-duty applications
  • Blue nitrile coating provides grip plus additional resistance to cuts and abrasions
  • Machine washable for longer service life
Compliance
ANSI™ Level 2 cut protection; EN388 4242
Specifications
Specifications
Smooth
Blue
HPPE (High-performance Polyethylene)
Light weight
Level 2
construction, recycling, lumber and mill work, Glass handling, Metal stamping and automotive assembly
Medium
Blue, White