Learn More
Honeywell™ Pure Fit™ Polyurethane-Coated Dyneema Gloves
Superior dexterity and tactile sensitivity
Manufacturer:  Honeywell™ PF540XL
 View more versions of this product
Catalog No. 19-816-642
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Lightweight Dyneema fabric with polyurethane palm and fingertip coating for increased cut, slash and abrasion resistance
  • Low lint for clean applications
  • Suited for use in electronic and small parts assembly, high-tech applications, automotive subassembly, and inspection
Compliance
ANSI™ Level 1 cut protection
Specifications
Specifications
Smooth
White
HPPE (High-performance Polyethylene)
Light weight
Level 2
protection and dexterity in applications requiring precision
X-Large
White