Honeywell Pure Fit™ Polyurethane-Coated Dyneema Gloves
Superior dexterity and tactile sensitivity
Manufacturer: Honeywell PF542L
 View more versions of this product
Catalog No. 19-148-222
  Check Availability
Add to Cart
    Specifications
    Hem Color Yellow
    For Use With (Application) electronic/small parts assembly, high tech applications, automotive sub assembly, light fabrication, micro engineering, packaging, inspection and light industrial
    Material HPPE, Lycra™
    Size Large
    Coating Material Smooth
    Color Gray
    Cut Resistance Rating Level 1
  • Description & Specifications
Related Products
Product Content Feedback