Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Ranger™ 1000 Boots

Offers complete protection and lightweight comfort

Manufacturer:  Total Fire Group BT1000095M

 View more versions of this product

Catalog No. 19-826-031


  Check Availability
Add to cart

Alert:

This product contains natural rubber latex proteins which may cause allergic reactions in some individuals.


Specifications Description & Specifications

Specifications

Steel
NFPA 1971, NFPA 1992, CSA and Mustard HD Protocol TOP-8-2-501
Natural and Butyl Rubber
Medium width
38.1cm
Steel
6.3 lb.
Black
9.5
Ideal to provide an insulation
15 in.
No
2.85kg
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit