Learn More
Honeywell Ranger™ 1500 Insulated Firefighting Boots, Narrow Width
Offers protection against punctures, moisture and electrical shocks
Manufacturer:  Total Fire Group BT1500040N
 View more versions of this product
Catalog No. 18-818-002
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Rubber; Kevlar/Nomex lining
NFPA Certified
7 lb.
4N
16 in.
Black/Yellow