Honeywell Ranger™ 1600 Insulated Firefighting Boots, Medium Width Available on GSA/VA Contract for Federal Government customers only.
Offers protection against punctures, moisture and electrical shocks
Manufacturer: Total Fire Group BT1600095M
 View more versions of this product
Catalog No. 18-818-104
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Weight (English) 7.1 lb.
    Height (English) 16 in.
    Color Black/Yellow
    Material Rubber; Wool Felt Lining
    Size 9.5M
    Certifications/Compliance NFPA 1971 and NFPA 1992
  • Description & Specifications
Related Products
Product Content Feedback