Learn More
Honeywell Ranger™ 2500 Noninsulated Firefighting Boots, Medium Width Available on GSA/VA Contract for Federal Government customers only.
Offers protection against punctures, moisture and electrical shocks
Manufacturer:  Total Fire Group BT2500055M
 View more versions of this product
Catalog No. 18-818-165
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Rubber; Kevlar Nomex Lining
NFPA 1971 and NFPA 1992
6.6 lb.
5.5M
16 in.
Black/Yellow