Honeywell Ranger™ 2500 Noninsulated Firefighting Boots, Medium Width Available on GSA/VA Contract for Federal Government customers only.
Offers protection against punctures, moisture and electrical shocks
Manufacturer: Total Fire Group BT2500140M
 View more versions of this product
Catalog No. 18-818-182
  Check Availability
Add to Cart
    Specifications
    Size 14M
    Material Rubber; Kevlar Nomex Lining
    Color Black/Yellow
    Height (English) 16 in.
    Weight (English) 6.6 lb.
    Certifications/Compliance NFPA 1971 and NFPA 1992
  • Description & Specifications
Related Products
Product Content Feedback