Learn More
Honeywell Ranger™ 2500 Noninsulated Firefighting Boots, Wide Width
Offers protection against punctures, moisture and electrical shocks
Manufacturer:  Total Fire Group BT2500060W
 View more versions of this product
Catalog No. 18-818-189
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
NFPA 1971 and 1992
6.6 lb.
Rubber; Kevlar Nomex Lining
16 in.
Black/Yellow
6W