Honeywell Ranger™ 3129 Shoe-Fit Women's Fire Boots - Wide Width
Wide-width fire boots with Shoe-Fit™ construction and Kevlar™ lining
Manufacturer: Total Fire Group 3129B8W
 View more versions of this product
Catalog No. 19-150-1392
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Certifications/Compliance NFPA 1971, 2007 Edition; Berry Amendment compliant
    Size 8W
    Material Urethane/Silicone Insulation; Kevlar Lining
    Height (English) 13 in.
  • Description & Specifications
Related Products
Product Content Feedback