Learn More
Honeywell™ Ranger™ 5104 Shoe-Fit Firewalker Boots
Work like a boot, but fit like a shoe
Manufacturer: Honeywell™ BT51049.5M
Description
Contouring snugs the foot into form-fitting pockets of support and comfort.
Standard Insulation Style 5104
- Wool felt linings and midsole
- Instep visibility band
- Extra heel reinforcement
- White Sure-Loc™ outsole
- Kevlar™/Nomex™ linings and wool felt midsole
- Built-in metatarsal foot guard and cushion shin protection
- Extra heel reinforcement
- Black Sure-Loc outsole
Order Info
Available in whole and half sizes from 5-15, in narrow (A-C), medium (D-E) or wide (2E-3E). To order sizes not listed here, contact your Fisher Safety Customer Service Representative.
Compliance
Meet NFPA 1971, 2007 Edition.
Specifications
NFPA 1971, 2007 Edition | |
16 in. | |
Rubber, Wool | |
9.5M |