Honeywell Saline Wall Station

The Honewell Saline Wall Station is easily mounted in hazardous areas to provide immediate relief in case of eye injury.

Manufacturer: Honeywell 320004620000

 View more versions of this product

Catalog No. 19-003-035

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

Since the first few seconds of an eye injury are critical, an eyewash bottle station will provide immediate relief until the worker has access to either a plumbed station or a shower. Eyewash bottles support a plumbed or self-contained drenching facility. They do not take the place of a 15-minute flushing facility.


    Specifications

    Product Type Double eye wall station
    Capacity (English) 32 oz.
    Capacity (Metric) 946mL
    Capacity 32 oz., 946mL
    Certifications/Compliance FDA
    For Use With (Application) Flushing nuisance particles such as pollen, dust, saw dust, smoke and more
    Includes Sperian Saline Buffered, Saline solution superior to tap Water
    Material Polyethylene
    Shelf Life 3 Years

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit