Honeywell Sensi-Task Natural Rubber Latex Gloves

Manufacturer: Honeywell T425/XL

 View all options for this product

Catalog No. 19-150-1029

  Check Availability
Add to Cart


    Coating Material Smooth
    Color White
    Disposable Yes
    Glove Cuff Style Rolled
    Hand Ambidextrous
    Grade Medical
    Length (English) 9 in.
    Length (Metric) 23cm
    Material Natural Rubber
    Powdered Yes
    Size X-Large
    Thickness 5mil
    Certifications USDA, FDA, 21 CFR

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit