Honeywell Sensi-Task Natural Rubber Latex Gloves

Manufacturer: Honeywell T425PF/L

 View more versions of this product

Catalog No. 19-150-1032

  Check Availability
Add to Cart

    Specifications

    Certifications USDA, FDA, 21 CFR
    Glove Cuff Style Rolled
    Hand Ambidextrous
    Grade Medical
    Length (English) 9 in.
    Length (Metric) 23cm
    Powdered No
    Color White
    Disposable Yes
    Material Natural Rubber
    Size Large
    Thickness 5mil
    Coating Material Smooth

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit