Honeywell Solutions for Eyesaline Porta-Stream Portable Eyewash Stations

Manufacturer: Honeywell 320005020000

 View more versions of this product

Catalog No. 17-292-3C

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Refill the eyewash device
    For Use With (Equipment) Porta Stream portable eyewash station
    Item Description Pre-mixed replacement saline solutions
    Volume (English) 128 oz
    Volume (Metric) 3785.41mL
    Product Type Saline Solution,Saline Solution
    Capacity (English) 128 oz.
    Capacity (Metric) 3785.41mL
    Capacity 128 oz., 3785.41mL
    Shelf Life 2.5 Years

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit