Honeywell Tails EWR Bronze BPR Structural Coats, Buckley Specs Available on GSA/VA Contract for Federal Government customers only.
Shortened front reduces garment weight and frees upper legs for unhampered climbing and crawling
Manufacturer: Total Fire Group GSABUC00112 SM
 View more versions of this product
Catalog No. 19-167-041A
  Check Availability
Add to Cart
    Specifications
    Size Small
    Material 7.5 oz. Gemini Matrix EWR
    Color Bronze
  • Description & Specifications
Related Products
Product Content Feedback