Honeywell Tails PBI Aluminized LTO Proximity Coats
Lighter weight with same protection, greater mobility, and reduced stress load
Manufacturer: Total Fire Group SAEGSA004772XL
 View all options for this product
Catalog No. 19-165-755
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Color Silver
    Size 2X-Large
    Material Aluminized PBI Silver; Crosstech/Nomex
    • Outer shell of 7 oz. aluminized PBI silver
    • Two-layer spunlace/meta-aramid thermal liner
    • Detachable inner liner with label pocket
    • CrossTech™/Nomex™ moisture barrier
    • Standard coat cuffs and Nomex-tabbed long wristlets
    • Exterior hook and D closures with 1 1/2 in. Velcro™ interior
    • Articulating rapid rescue strap
    With Pockets
    • Half hi bellows pockets of aluminized PBI silver: 6 x 9 x 1 1/2 in.
    • Standard chinstrap
    • Dead air panels
    • Two take-up postman straps
    • Navy NFPA 2007 Edition specifications
    Without Pockets
    • Inspection port liner
    • Extended dead air panels
    • Standard coat cuffs and comfort chinstrap of aluminized PBI silver
    • Marine Corp NFPA 2007 Edition specifications
  • Description & Specifications
Related Products
Product Content Feedback