Honeywell Tails PBI Aluminized Silver BPR Proximity Coats, Goodfellow Specs
Outer shell of 7 oz. aluminized PBI silver provides excellent radiant heat reflectivity for firefighting applications of extreme heat
Manufacturer: Total Fire Group GSAGOO00533 MD
 View more versions of this product
Catalog No. 19-167-049B
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Size Medium
    Material 7 oz. Aluminized PBI Silver
    Color Silver
  • Description & Specifications
Related Products
Product Content Feedback