Honeywell Tails PBI Aluminized Silver BPR Proximity Pants, Buckley Specs
Outer shell of 7 oz. aluminized PBI silver provides excellent radiant heat reflectivity for firefighting applications of extreme heat
Manufacturer: Total Fire Group GSABUC00111LG
 View more versions of this product
Catalog No. 19-167-040C
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Color Silver
    Size Large
    Material 7 oz. Pbi/Kevlar
    • Outer shell of 7 oz. aluminized PBI silver
    • Two-layer spunlace/meta-aramid facecloth thermal liner
    • Crosstech™/Nomex™ facecloth moisture barrier
    • 1.5 in. Velcro™ fly
    • Inspection port liner
    • Detachable liner
    • Angled cuffs
    • Replaceable heat-channel knees with military-style frames
    • Two take-up postman straps
    • Bellows pockets measuring 9 x 9 x 1.5 in., with right pocket divided with PBI EWR bronze divider 3.5 in. from rear
    • Installed Dynaback™ suspenders
    • NFPA 2007 Edition Specifications
  • Description & Specifications
Related Products
Product Content Feedback