Honeywell Tails PBI Aluminized Silver BPR Proximity Pants, Goodfellow Specs
Outer shell of 7 oz. aluminized PBI silver provides excellent radiant heat reflectivity for firefighting applications of extreme heat
Manufacturer: Total Fire Group GSAGOO00534XXL
 View more versions of this product
Catalog No. 19-167-050E
  Check Availability
Add to Cart
    Specifications
    Size 2X-Large
    Material 7 oz. Pbi/Kevlar
    Color Silver
  • Description & Specifications
Related Products
Product Content Feedback