Honeywell Tails PBI Aluminized Silver LTO Proximity Coats, Navy Specs
Outer shell of 7 oz. aluminized PBI silver provides excellent radiant heat reflectivity for firefighting applications of extreme heat
Manufacturer: Total Fire Group GSANAY00213XL
 View more versions of this product
Catalog No. 19-167-037D
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Size X-Large
    Material 7 oz. Aluminum Pbi Shell; Crosstech/Nomex
    Color Silver
  • Description & Specifications
Related Products
Product Content Feedback