Learn More
Honeywell Tuff-Coat Cut-Resistant Gloves
Seamless knit liners with durable quality coatings
Manufacturer: Honeywell KV200M
Description
Ideal for construction, forestry, municipal services, and transportation.
- Standard weight, 7-gauge Kevlar seamless knit shell
- Kevlar provides outstanding cut and slash protection
- Natural rubber palm and fingertip coating
- Rubber coating provides excellent grip and flexibility
- This product contains natural rubber
Compliance
Meets ANSI A2 (cut level) standards.
Specifications
Knit | |
construction, forestry, municipal services and transportation | |
Medium | |
Blue, Yellow |
Blue | |
Dupont™ Kevlar™ Aramid Fiber | |
Medium weight |